Learn More
Aviva Systems Biology Corp Esrp2 Antibody - C-terminal region (ARP42490_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42490_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Esrp2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 9530027K23Rik, Rbm35bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-005