Learn More
Aviva Systems Biology Corp ESRRG Antibody - N-terminal region (ARP45602_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45602_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ESRRG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ESRRG. Alternate names: DKFZp781L1617, ERR3, FLJ16023, KIAA0832, NR3B3, ERRgammaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59062