Learn More
Aviva Systems Biology Corp ESSPL Antibody - N-terminal region (ARP58620_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58620_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ESSPL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ESSPL. Alternate names: ESSPLApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Pig, Rabbit, Rat
Catalog No. 50-014-62377