Learn More
Medchemexpress LLC HY-131914A 5mg Medchemexpress, EST64454 (hydrochloride) CAS:1950569-11-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY131914A5MG
Encompass_Preferred
Medchemexpress, HY-131914A 5mg EST64454 (hydrochloride) CAS:1950569-11-5 EST64454 hydrochloride is a selective and orally active sigma-1 receptor antagonist with a Ki of 22 nM. EST64454 hydrochloride has the potential for the research of the pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1299