Learn More
Medchemexpress LLC HY-134189A 5mg Medchemexpress, EST73502 (hydrochloride) CAS:2535970-65-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY134189A5MG
Encompass_Preferred
Medchemexpress, HY-134189A 5mg EST73502 (hydrochloride) CAS:2535970-65-9 EST73502 hydrochloride is a selective, orally active and blood-brain barrier (BBB) penetrant dual μ-opioid receptor (MOR) agonist and σ1 receptor (σ1R) antagonist, with Kis of 64 nM and 118 nM for MOR and σ1R, respectively. EST73502 hydrochloride has antinociceptive activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1358