Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Est73502 monohydrochloride | 2535970-65-9 | 99.6% | C19H27ClF2N2O2 | 10MG
SDP

Catalog No. 5000258583
Encompass_Preferred

EST73502 monohydrochloride is the hydrochloride salt of EST73502, a research chemical described as a selective, orally active, blood-brain barrier penetrant dual μ-opioid receptor agonist and σ1 receptor antagonist. It is provided for laboratory research use; consult the supplier datasheet and safety data sheet for handling, storage, and certificate of analysis details.

  • Hydrochloride salt form for improved solubility.
  • Reported molecular formula C19H27ClF2N2O2.
  • Molecular weight 388.88 g/mol.
  • High reported purity in distributor listings; verify with certificate of analysis.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-58583 Supplier Medchemexpress LLC Supplier No. HY134189A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EST73502 monohydrochloride is the hydrochloride salt of EST73502, a research chemical described as a selective, orally active, blood-brain barrier penetrant dual μ-opioid receptor agonist and σ1 receptor antagonist. It is provided for laboratory research use; consult the supplier datasheet and safety data sheet for handling, storage, and certificate of analysis details.

  • Hydrochloride salt form for improved solubility.
  • Reported molecular formula C19H27ClF2N2O2.
  • Molecular weight 388.88 g/mol.
  • High reported purity in distributor listings; verify with certificate of analysis.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.