Learn More
Genscript Corporation eStain Gel Holder
Catalog No. NC1456034
Encompass
GenScript Rabbit Anti-E-tag [HRP] Polyclonal Antibody is highly purified from rabbit antiserum by immunoaffinity chromatography and then conjugated to horseradish peroxidase.GenScript Rabbit Anti-E-tag [HRP] Polyclonal Antibody is suitable for detecting fusion proteins that contain E-tags.
Catalog No. NC1456034 Supplier Genscript Corporation Supplier No. L00658
GenScript Rabbit Anti-E-tag [HRP] Polyclonal Antibody is highly purified from rabbit antiserum by immunoaffinity chromatography and then conjugated to horseradish peroxidase.GenScript Rabbit Anti-E-tag [HRP] Polyclonal Antibody is suitable for detecting fusion proteins that contain E-tags.