Learn More
Genscript Corporation eStain L1C Protein Staining Kit
Catalog No. NC1630149
Encompass
eStain L1C Staining Kit (Cat No: L00753) contains reagents that are used with eStain L1 protein staining device (L00657). Each kit contains 1 x 2L of staining solution, 1 x 2L of destaining solution, and a pack of filter paper (50pK).
Catalog No. NC1630149 Supplier Genscript Corporation Supplier No. L00753
eStain L1C Staining Kit (Cat No: L00753) contains reagents that are used with eStain L1 protein staining device (L00657). Each kit contains 1 x 2L of staining solution, 1 x 2L of destaining solution, and a pack of filter paper (50pK).