Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardinal Health™ Esteem™ Comfort Nitrile Gloves

Catalog No. 19162470 Shop All Cardinal Health Products
Change view
Click to view available options
Size:
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-162-470 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Caution: Gloves used for protection against chemotherapy drug and laboratory chemical exposure should be selected specifically for the type of chemicals being used. Review Safety Data Sheets (SDS) for the chemicals/drugs being used to determine required level of protection.
Catalog No. 19-162-470 Supplier Cardinal Health™ Supplier No. 88ES02S
This item is not returnable. View return policy
Caution: Gloves used for protection against chemotherapy drug and laboratory chemical exposure should be selected specifically for the type of chemicals being used. Review Safety Data Sheets (SDS) for the chemicals/drugs being used to determine required level of protection.

Stretchy nitrile exam gloves feature a 30% higher elongation than the minimum ASTM standard.

  • Stretchy nitrile material
  • Textured fingertips provide a secure grip, even when wet
  • Beaded cuffs reduce tears during donning
  • Powder free
  • non-sterile
  • Latex-free

Specifications

Certifications/Compliance D6319, D6978, and F739 ASTM Standards
Chemotherapeutic Rated Yes
Applications Healthcare, Laboratory
AQL 1
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 4.3 mil
Thickness (Metric) 0.10 mm
FDA 510 (k) No
Model 88ES02S
Series Esteem™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.