Learn More
MSC SC Johnson Professional Estesol PURE Cleaner, Lotion, 2L, 4/Case
Catalog No. 502868683
Encompass
Estesol PURE Hand, Hair & Body Cleaner is a fragrance-free, dye-free, skin hypoallergenic lotion designed for general industrial and food manufacturing/processing environments. It effectively removes light duty oils, greases, general grime, and food-related soilings, and is also suitable for industrial shower environments. Formulated with a low pH value (4.0-6.5), it helps minimize disruption to the skin's acid mantle and contains glycerin to prevent dryness.
  • Ideal for high frequency use
  • Formulated with a low pH value (4.0-6.5)
  • Skin hypoallergenic with a mild preservative
  • Silicone-free formulation
  • Contains glycerin to moisturize skin
  • Suitable for food facilities
  • Meets NSF, Green Seal, UL ECOLOGO, and USDA Certified Biobased Product standards
Catalog No. 50-286-8683 Supplier MSC Supplier No. 44346310
Estesol PURE Hand, Hair & Body Cleaner is a fragrance-free, dye-free, skin hypoallergenic lotion designed for general industrial and food manufacturing/processing environments. It effectively removes light duty oils, greases, general grime, and food-related soilings, and is also suitable for industrial shower environments. Formulated with a low pH value (4.0-6.5), it helps minimize disruption to the skin's acid mantle and contains glycerin to prevent dryness.
  • Ideal for high frequency use
  • Formulated with a low pH value (4.0-6.5)
  • Skin hypoallergenic with a mild preservative
  • Silicone-free formulation
  • Contains glycerin to moisturize skin
  • Suitable for food facilities
  • Meets NSF, Green Seal, UL ECOLOGO, and USDA Certified Biobased Product standards