Learn More
Medchemexpress LLC Estra-1,3,5(10)-trien-17-one, 3-[(aminosulfonyl)oxy]- | 148672-09-7 | 99.99% | C18H23NO4S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1458550MG
Encompass_Preferred
Estrone O-sulfamate is a potent steroid sulfatase (STS) inhibitor. It exhibits inhibitory activity for STS in placental microsomes preparation and in MCF-7 cells, and can be utilized for cancer research.
  • Potent steroid sulfatase (STS) inhibitor
  • Exhibits inhibitory activity for STS in placental microsomes and MCF-7 cells
  • Can be used for cancer research
  • Shows inhibitory activity against human steroid sulfatase over-expressed in CHO cells
  • Exhibits antiproliferative activity against various human cancer cell lines
  • Inhibits human placental steroid sulfatase in HEK293 cells
  • Inhibits estrone sulfatase in JEG-3 cells
  • Causes growth inhibition and antiproliferative activity in MCF7 cells
Catalog No. 50-003-55948