Learn More
Medchemexpress LLC Estra-1,3,5(10)-trien-17-one, 3-hydroxy-, O-(carboxymethyl)oxime, (17E)- | 693227-00-8 | 99.5% | 343.42 | 250 MG
SDP
Catalog No. 5000402836
Encompass_Preferred
Androgen receptor ligand 3 is an androgen receptor (AR) ligand. It is used in the formation of an ERα PROTAC degrader by linking to an IAP ligand via a linker.
  • Acts as an androgen receptor ligand.
  • Utilized in ERα PROTAC degrader formation.
Catalog No. 50-004-02836 Supplier Medchemexpress LLC Supplier No. 250MGHY43962
Androgen receptor ligand 3 is an androgen receptor (AR) ligand. It is used in the formation of an ERα PROTAC degrader by linking to an IAP ligand via a linker.
  • Acts as an androgen receptor ligand.
  • Utilized in ERα PROTAC degrader formation.