Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estra-1,3,5(10)-trien-17-one, 3-(sulfooxy)-, compd. with piperazine (1:1) | 7280-37-7 | 98.7% | 1 ML
SDP

Catalog No. 5000412518
Encompass_Preferred

Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.

  • CAS number: 7280-37-7
  • Molecular formula: C₂₂H₃₂N₂O₅S
  • Molecular weight: 436.56
  • Target: Estrogen Receptor/ERR
  • Pathway: Vitamin D Related/Nuclear Receptor
  • Storage (solid): 4°C, sealed storage, away from moisture
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Appearance: White to off-white solid
  • Purity: 98.72%
  • In vitro solubility (DMSO): 110 mg/mL (251.97 mM)

Catalog No. 50-004-12518 Supplier Medchemexpress LLC Supplier No. HYB136110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estropipate is a form of estrogen, used to treat symptoms of menopause, also used to prevent osteoporosis.

  • CAS number: 7280-37-7
  • Molecular formula: C₂₂H₃₂N₂O₅S
  • Molecular weight: 436.56
  • Target: Estrogen Receptor/ERR
  • Pathway: Vitamin D Related/Nuclear Receptor
  • Storage (solid): 4°C, sealed storage, away from moisture
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture)
  • Appearance: White to off-white solid
  • Purity: 98.72%
  • In vitro solubility (DMSO): 110 mg/mL (251.97 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.