Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estra-1,3,5(10)-triene-2,4,16,16-d4-3,17-diol, 3-(hydrogen sulfate), sodium salt, (17a)- | 352431-50-6 | 99.6% | 378.45 g/mol | C18H19D4NaO5S | 5 MG
SDP

Catalog No. 5000220989
Encompass_Preferred

17β-Estradiol sulfate-d4 (sodium) is a deuterium-labeled sodium salt of 17β-estradiol 3-sulfate supplied as a high-purity analytical standard for research use. It is intended as an internal standard or tracer in mass spectrometry and other quantitative assays to study estradiol metabolism and sulfate conjugates.

  • Deuterium-labeled (d4) analog for isotope dilution and tracer studies.
  • Sodium salt supplied as a solid, suitable for long-term storage.
  • High purity (~99.6%) for analytical applications.
  • Molecular formula C18H19D4NaO5S; molecular weight 378.45 g/mol.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-20989 Supplier Medchemexpress LLC Supplier No. HY141672S15MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

17β-Estradiol sulfate-d4 (sodium) is a deuterium-labeled sodium salt of 17β-estradiol 3-sulfate supplied as a high-purity analytical standard for research use. It is intended as an internal standard or tracer in mass spectrometry and other quantitative assays to study estradiol metabolism and sulfate conjugates.

  • Deuterium-labeled (d4) analog for isotope dilution and tracer studies.
  • Sodium salt supplied as a solid, suitable for long-term storage.
  • High purity (~99.6%) for analytical applications.
  • Molecular formula C18H19D4NaO5S; molecular weight 378.45 g/mol.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.