Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estra-1,3,5(10)-triene-3,17-diol (17β)-, 17-(hydrogen butanedioate) | 7698-93-3 | 98.8% | 372.45 | 100 MG
SDP

Catalog No. 5000360980
Encompass_Preferred

β-Estradiol 17-hemisuccinate is a synthetic derivative of estradiol. This compound is a white to off-white solid with a purity of 98.8%. It is suitable for research applications.

  • Synthetic derivative of estradiol
  • Purity of 98.8%
  • Molecular weight of 372.45
  • White to off-white solid appearance
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO

Catalog No. 50-003-60980 Supplier Medchemexpress LLC Supplier No. HY148586100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Estradiol 17-hemisuccinate is a synthetic derivative of estradiol. This compound is a white to off-white solid with a purity of 98.8%. It is suitable for research applications.

  • Synthetic derivative of estradiol
  • Purity of 98.8%
  • Molecular weight of 372.45
  • White to off-white solid appearance
  • Store at 4°C under nitrogen
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.