Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estra-1,3,5(10)-triene-3,17-diol (17β)-, 17-(hydrogen butanedioate) | 7698-93-3 | 98.8% | 372.45 | 50 MG
SDP

Catalog No. 5000360627
Encompass_Preferred

β-Estradiol 17-hemisuccinate is a synthetic derivative of estradiol, appearing as a white to off-white solid.

  • Synthetic derivative of estradiol.
  • CAS number: 7698-93-3.
  • Molecular weight: 372.45.
  • Chemical formula: C22H28O5.
  • Purity: 98.8%.
  • Store at 4°C, stored under nitrogen.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, stored under nitrogen.
  • 1H NMR spectrum and LCMS consistent with structure.

Catalog No. 50-003-60627 Supplier Medchemexpress LLC Supplier No. HY14858650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

β-Estradiol 17-hemisuccinate is a synthetic derivative of estradiol, appearing as a white to off-white solid.

  • Synthetic derivative of estradiol.
  • CAS number: 7698-93-3.
  • Molecular weight: 372.45.
  • Chemical formula: C22H28O5.
  • Purity: 98.8%.
  • Store at 4°C, stored under nitrogen.
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, stored under nitrogen.
  • 1H NMR spectrum and LCMS consistent with structure.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.