Learn More
Medchemexpress LLC Estra-1,3,5(10)-triene-3,17-diol (17β)-, 3-sulfamate | 172377-52-5 | MFCD00947822 | 351.46 | 25 MG
SDP
Catalog No. 5000449889
Encompass_Preferred
Estradiol 3-sulfamate, also identified as BLE 00084, E2MATE, or ES-J 995, is characterized as a potent, long-acting, and orally active inhibitor of steroid sulfatase.
  • Potent steroid sulfatase inhibitor
  • Long-acting steroid sulfatase inhibitor
  • Orally active steroid sulfatase inhibitor
  • Inhibits estrone sulfatase with an IC50 of 251 nM
  • Inhibits estrone sulfatase with a Ki of 133 nM
Catalog No. 50-004-49889 Supplier Medchemexpress LLC Supplier No. 25MGHYU00112
Estradiol 3-sulfamate, also identified as BLE 00084, E2MATE, or ES-J 995, is characterized as a potent, long-acting, and orally active inhibitor of steroid sulfatase.
  • Potent steroid sulfatase inhibitor
  • Long-acting steroid sulfatase inhibitor
  • Orally active steroid sulfatase inhibitor
  • Inhibits estrone sulfatase with an IC50 of 251 nM
  • Inhibits estrone sulfatase with a Ki of 133 nM