Learn More
Medchemexpress LLC Estradiol-13C2 (β-estradiol-13C2) | 82938-05-4 | 1 MG
SDP
Catalog No. 5000450898
Encompass_Preferred
Estradiol-13C2 is the 13C-labeled version of Estradiol, which is a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. Estradiol also upregulates IL-6 expression via the estrogen receptor β (ERβ) pathway.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • It is a 13C-labeled compound, indicating it is a stable isotope
Catalog No. 50-004-50898 Supplier Medchemexpress LLC Supplier No. HYB0141S51MG
Estradiol-13C2 is the 13C-labeled version of Estradiol, which is a steroid sex hormone crucial for maintaining fertility and secondary sexual characteristics in females. Estradiol also upregulates IL-6 expression via the estrogen receptor β (ERβ) pathway.
  • Used as a tracer
  • Serves as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS
  • It is a 13C-labeled compound, indicating it is a stable isotope