Learn More
Medchemexpress LLC β-Estradiol 17-acetate | 1743-60-8 | 99.9% | 1 ML
SDP
Catalog No. 5000406749
Encompass_Preferred
β-Estradiol 17-acetate is a metabolite of estradiol, designed for research use only in laboratory settings. It is provided for chemical synthesis and intended for use by experienced personnel in appropriately equipped facilities.
  • Available in powder and pre-dissolved solution formats.
  • Stable under recommended storage conditions, with specific guidance for powder and solvent forms.
  • Highly soluble in DMSO for in vitro applications.
  • Features established protocols for in vivo solubility using various solvent systems.
  • Molecular formula C20H26O3 and molecular weight of 314.42.
  • Typically appears as a white to off-white solid with a melting point of 119°C.
Catalog No. 50-004-06749 Supplier Medchemexpress LLC Supplier No. HYB070810MM/1ML
β-Estradiol 17-acetate is a metabolite of estradiol, designed for research use only in laboratory settings. It is provided for chemical synthesis and intended for use by experienced personnel in appropriately equipped facilities.
  • Available in powder and pre-dissolved solution formats.
  • Stable under recommended storage conditions, with specific guidance for powder and solvent forms.
  • Highly soluble in DMSO for in vitro applications.
  • Features established protocols for in vivo solubility using various solvent systems.
  • Molecular formula C20H26O3 and molecular weight of 314.42.
  • Typically appears as a white to off-white solid with a melting point of 119°C.