Learn More
Medchemexpress LLC Estradiol 17-(β-D)-glucuronide | 1806-98-0 | MFCD00198971 | 99.0% | 448.51 g/mol | C24H32O8 | 1 MG
SDP
Catalog No. 5000285141
Encompass_Preferred
Estradiol 17-(β-D)-glucuronide is a glucuronidated metabolite of the estrogen estradiol used as a research reagent. It functions as a cholestatic agent and a high-affinity substrate for OATP organic anion transporters, and is commonly applied in studies of hepatic transport, bile acid homeostasis, and estrogen metabolism.
  • Used as a cholestatic agent in in vitro and in vivo studies.
  • High-affinity substrate for OATP organic anion transporters.
  • Provided as a high-purity research-grade compound (purity 99.0%).
  • Supplied in milligram quantities suitable for biochemical assays.
  • Molecular formula C24H32O8; molecular weight 448.51 g/mol.
Catalog No. 50-002-85141 Supplier Medchemexpress LLC Supplier No. HYW3531021MG
Add to Cart
Add to Cart
Estradiol 17-(β-D)-glucuronide is a glucuronidated metabolite of the estrogen estradiol used as a research reagent. It functions as a cholestatic agent and a high-affinity substrate for OATP organic anion transporters, and is commonly applied in studies of hepatic transport, bile acid homeostasis, and estrogen metabolism.
  • Used as a cholestatic agent in in vitro and in vivo studies.
  • High-affinity substrate for OATP organic anion transporters.
  • Provided as a high-purity research-grade compound (purity 99.0%).
  • Supplied in milligram quantities suitable for biochemical assays.
  • Molecular formula C24H32O8; molecular weight 448.51 g/mol.