Learn More
Medchemexpress LLC Estradiol 17-(β-D-glucuronide) (sodium) | 15087-02-2 | 98.9% | 25 MG
SDP
Catalog No. 5000339178
Encompass_Preferred
Estradiol 17-(β-D-Glucuronide) sodium is a D-ring glucuronide metabolite of natural estrogen formed in the liver. It acts as a substrate for the organic anion-transporting polypeptide family (Oatp) and multidrug resistance-associated protein 2 (Mrp2). This compound regulates MRP8-mediated transport processes and is suitable for studies related to intrahepatic cholestasis.
  • D-ring glucuronide metabolite of natural estrogen
  • Substrate for organic anion-transporting polypeptide family (Oatp) and multidrug resistance-associated protein 2 (Mrp2)
  • Regulates MRP8-mediated transport processes
  • Inhibits MRP8-mediated transport of dehydroepiandrosterone 3-sulfate and taurocholic acid
  • Induces immediate, reversible reduction of bile flow
  • Suitable for studies related to intrahepatic cholestasis
Catalog No. 50-003-39178 Supplier Medchemexpress LLC Supplier No. 25MGHY129987
Estradiol 17-(β-D-Glucuronide) sodium is a D-ring glucuronide metabolite of natural estrogen formed in the liver. It acts as a substrate for the organic anion-transporting polypeptide family (Oatp) and multidrug resistance-associated protein 2 (Mrp2). This compound regulates MRP8-mediated transport processes and is suitable for studies related to intrahepatic cholestasis.
  • D-ring glucuronide metabolite of natural estrogen
  • Substrate for organic anion-transporting polypeptide family (Oatp) and multidrug resistance-associated protein 2 (Mrp2)
  • Regulates MRP8-mediated transport processes
  • Inhibits MRP8-mediated transport of dehydroepiandrosterone 3-sulfate and taurocholic acid
  • Induces immediate, reversible reduction of bile flow
  • Suitable for studies related to intrahepatic cholestasis