Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estradiol 17-(β-D-Glucuronide) sodium | 15087-02-2 | 98.9% | 50 MG
SDP

Catalog No. 5000338545
Encompass_Preferred

Estradiol 17-(β-D-Glucuronide) sodium is a D-ring glucuronide metabolite of natural estrogen formed in the liver. It functions as a substrate of the organic anion-transporting polypeptide family (Oatp) and multidrug resistance-associated protein 2 (Mrp2). This compound regulates and inhibits MRP8-mediated transport processes and can be used in studies related to intrahepatic cholestasis, as it induces immediate, reversible reduction of bile flow and acute intrahepatic cholestasis in female rats.

  • Metabolite of natural estrogen
  • Substrate of Oatp and Mrp2
  • Regulates and inhibits MRP8-mediated transport processes
  • Induces immediate, reversible reduction of bile flow and acute intrahepatic cholestasis
  • Useful in studies related to intrahepatic cholestasis
  • Solid, white to off-white appearance
  • Molecular weight of 470.49
  • Chemical formula C24H31NaO8

Catalog No. 50-003-38545 Supplier Medchemexpress LLC Supplier No. HY12998750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estradiol 17-(β-D-Glucuronide) sodium is a D-ring glucuronide metabolite of natural estrogen formed in the liver. It functions as a substrate of the organic anion-transporting polypeptide family (Oatp) and multidrug resistance-associated protein 2 (Mrp2). This compound regulates and inhibits MRP8-mediated transport processes and can be used in studies related to intrahepatic cholestasis, as it induces immediate, reversible reduction of bile flow and acute intrahepatic cholestasis in female rats.

  • Metabolite of natural estrogen
  • Substrate of Oatp and Mrp2
  • Regulates and inhibits MRP8-mediated transport processes
  • Induces immediate, reversible reduction of bile flow and acute intrahepatic cholestasis
  • Useful in studies related to intrahepatic cholestasis
  • Solid, white to off-white appearance
  • Molecular weight of 470.49
  • Chemical formula C24H31NaO8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.