Learn More
Medchemexpress LLC Estradiol 17-(β-D-glucuronide) sodium | 15087-02-2 | MFCD00056557 | 99.5% | C24H31NaO8 | 10MG
SDP
Catalog No. 5000258847
Encompass_Preferred
Estradiol 17-(β-D-glucuronide) sodium is the sodium salt of a conjugated estrogen (E217G) formed in the liver and excreted in bile. It is used as a research reagent to investigate estrogen metabolism, transporter activity, and mechanisms of cholestasis. Supplied as a high-purity solid for in vitro and biochemical assays; for research use only.
  • High purity suitable for analytical and biochemical assays.
  • Serves as a substrate for hepatic transporters involved in bile excretion.
  • Relevant for studying intrahepatic cholestasis and estrogen metabolism.
  • Soluble in polar organic solvents such as DMSO and DMF.
  • Available in multiple small-scale pack sizes for lab use.
Catalog No. 50-002-58847 Supplier Medchemexpress LLC Supplier No. HY12998710MG
Estradiol 17-(β-D-glucuronide) sodium is the sodium salt of a conjugated estrogen (E217G) formed in the liver and excreted in bile. It is used as a research reagent to investigate estrogen metabolism, transporter activity, and mechanisms of cholestasis. Supplied as a high-purity solid for in vitro and biochemical assays; for research use only.
  • High purity suitable for analytical and biochemical assays.
  • Serves as a substrate for hepatic transporters involved in bile excretion.
  • Relevant for studying intrahepatic cholestasis and estrogen metabolism.
  • Soluble in polar organic solvents such as DMSO and DMF.
  • Available in multiple small-scale pack sizes for lab use.