Learn More
Cayman Chemical Estradiol 17-(β-D-Glucuronide) (sodium salt hydrate), Purity: ≥95%, 50 mg
SDP
Catalog No. 502970378
Encompass_Preferred
An estrogen metabolite that acts as a substrate of MRP2 (Km = 75 µM), and through MRP2-mediated transport, functions as a cholestatic agent, decreasing bile flow
Catalog No. 50-297-0378 Supplier Cayman Chemical Supplier No. 1615650 MG
An estrogen metabolite that acts as a substrate of MRP2 (Km = 75 µM), and through MRP2-mediated transport, functions as a cholestatic agent, decreasing bile flow