Learn More
Cayman Chemical Estradiol 17-(β-D-Glucuronide) (sodium salt hydrate), Purity: ≥95%, 5 mg
SDP
Catalog No. 502971826
Encompass_Preferred
An estrogen metabolite that acts as a substrate of MRP2 (Km = 75 µM), and through MRP2-mediated transport, functions as a cholestatic agent, decreasing bile flow
Catalog No. 50-297-1826 Supplier Cayman Chemical Supplier No. 161565 MG
An estrogen metabolite that acts as a substrate of MRP2 (Km = 75 µM), and through MRP2-mediated transport, functions as a cholestatic agent, decreasing bile flow