Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-Estradiol 17-hemisuccinate | 7698-93-3 | 98.8% | 372.45 | 1 MG
SDP

Catalog No. 5000360453
Encompass_Preferred

β-Estradiol 17-hemisuccinate is a synthetic derivative of estradiol, classified as a drug derivative. It is intended for research applications.

  • Synthetic derivative of estradiol
  • Molecular formula: C22H28O5
  • Molecular weight: 372.45
  • Purity: 98.83% (LCMS)
  • Appearance: White to off-white solid
  • Storage: 4°C, stored under nitrogen
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Catalog No. 50-003-60453 Supplier Medchemexpress LLC Supplier No. HY1485861MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Estradiol 17-hemisuccinate is a synthetic derivative of estradiol, classified as a drug derivative. It is intended for research applications.

  • Synthetic derivative of estradiol
  • Molecular formula: C22H28O5
  • Molecular weight: 372.45
  • Purity: 98.83% (LCMS)
  • Appearance: White to off-white solid
  • Storage: 4°C, stored under nitrogen
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month (stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.