Learn More
Cayman Chemical β-Estradiol 17-valerate, Purity: ≥98%, CAS: 979-32-8, 5 g
SDP
Catalog No. 502987850
Encompass_Preferred
A synthetic estrogen; increases ERβ, but not ERα, levels in the hippocampus and cortex of ovariectomized rats at 0.3 mg/day; Intradermal administration induces transient permanent diestrus and decreases litter size in rats at 5 µg/animal; has been used to induce PCOS in rats; has been found in wastewater effluent
Catalog No. 50-298-7850 Supplier Cayman Chemical Supplier No. 222215 G
A synthetic estrogen; increases ERβ, but not ERα, levels in the hippocampus and cortex of ovariectomized rats at 0.3 mg/day; Intradermal administration induces transient permanent diestrus and decreases litter size in rats at 5 µg/animal; has been used to induce PCOS in rats; has been found in wastewater effluent