Learn More
Apexbio Technology LLC Estradiol 50-28-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062385
Encompass_Preferred
Estradiol is an endogenous steroid hormone involved in the regulation of reproductive biology and cellular physiology through interaction with estrogen receptor (ER) subtypes and Estradiol binding to ER initiates genomic and non-genomic signaling pathways influencing transcription of diverse target genes related to proliferation differentiation and metabolism Estradiol-mediated ER signaling modulates cellular processes including epithelial-mesenchymal transition endothelial function oxidative stress responses and gene expression profiles in various cell lines such as HEK293 and U2OS Additionally estradiol treatment regulates cardiovascular function by inducing mitochondrial superoxide dismutase (SOD2) implicating protective effects in vascular conditions Estradiol exhibits ER subtype selectivity with reported ER and ER binding affinities (IC50) around 0 2-0 5 nM utilized broadly in estrogen signaling research and cardiovascular disease modelling
Catalog No. 50-000-62385 Supplier Apexbio Technology LLC Supplier No. A84255.1
Estradiol is an endogenous steroid hormone involved in the regulation of reproductive biology and cellular physiology through interaction with estrogen receptor (ER) subtypes and Estradiol binding to ER initiates genomic and non-genomic signaling pathways influencing transcription of diverse target genes related to proliferation differentiation and metabolism Estradiol-mediated ER signaling modulates cellular processes including epithelial-mesenchymal transition endothelial function oxidative stress responses and gene expression profiles in various cell lines such as HEK293 and U2OS Additionally estradiol treatment regulates cardiovascular function by inducing mitochondrial superoxide dismutase (SOD2) implicating protective effects in vascular conditions Estradiol exhibits ER subtype selectivity with reported ER and ER binding affinities (IC50) around 0 2-0 5 nM utilized broadly in estrogen signaling research and cardiovascular disease modelling