Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Estradiol-3-Benzoate, 99.7%, MP Biomedicals™

Catalog No. ICN19041105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19041105 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19041105 Supplier MP Biomedicals, Inc Supplier No. 0219041105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in a study of general steroid metabolism and clearing, as groundwork for anabolic steroid screening in athletes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-50-0
Molecular Formula C25H28O3
Molecular Weight (g/mol) 376.496
MDL Number MFCD00003692
InChI Key UYIFTLBWAOGQBI-BZDYCCQFSA-N
Synonym estradiol benzoate, estradiol monobenzoate, benzhormovarine, benzoestrofol, benovocylin, benzofoline, follidrin, ovasterol-b, benzo-gynoestryl, primogyn b
PubChem CID 222757
ChEBI CHEBI:77006
IUPAC Name [(8R,9S,13S,14S,17S)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] benzoate
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)OC(=O)C5=CC=CC=C5

Specifications

Color White
Assay Percent Range 99.7%
Quantity 5 g
Formula Weight 376.5
Physical Form Powder
Chemical Name or Material β-Estradiol-3-Benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.