Learn More
Cayman Chemical Estradiol 3-(β-D-Glucuronide) (sodium salt), Purity: ≥95%, CAS: 14982-12-8, 25 mg
SDP
Catalog No. 502970279
Encompass_Preferred
A non-cholestatic regioisomer of E217G that acts as a substrate for MRP2, competing with E217G for MRP2-mediated transport (IC50 = 14.2 µM)
Catalog No. 50-297-0279 Supplier Cayman Chemical Supplier No. 1615525 MG
A non-cholestatic regioisomer of E217G that acts as a substrate for MRP2, competing with E217G for MRP2-mediated transport (IC50 = 14.2 µM)