Learn More
Medchemexpress LLC Estradiol 3-glucuronide | 15270-30-1 | 97.6% | 448.51 g/mol | C24H32O8 | 500 UG
SDP
Catalog No. 5000284430
Encompass_Preferred
Estradiol 3-glucuronide is the glucuronic acid conjugate of estradiol used as a research reagent and immunogen for studies of estrogen metabolism, antibody production, and radioimmunoassay. Supplied as a white to off-white solid for laboratory research use only.
  • White to off-white solid suitable for biochemical assays.
  • Used as an immunogen for antibody generation and radioimmunoassay studies.
  • Applicable in research on estrogen metabolism and analytical detection methods.
  • Typical purity approximately 97.6% by HPLC (batch dependent).
  • Chemical formula C24H32O8; molecular weight 448.51 g/mol; CAS 15270-30-1.
  • Store powder at -20°C; in solvent store at -80°C up to 6 months or -20°C for shorter term.
Catalog No. 50-002-84430 Supplier Medchemexpress LLC Supplier No. HYW585842500UG
Estradiol 3-glucuronide is the glucuronic acid conjugate of estradiol used as a research reagent and immunogen for studies of estrogen metabolism, antibody production, and radioimmunoassay. Supplied as a white to off-white solid for laboratory research use only.
  • White to off-white solid suitable for biochemical assays.
  • Used as an immunogen for antibody generation and radioimmunoassay studies.
  • Applicable in research on estrogen metabolism and analytical detection methods.
  • Typical purity approximately 97.6% by HPLC (batch dependent).
  • Chemical formula C24H32O8; molecular weight 448.51 g/mol; CAS 15270-30-1.
  • Store powder at -20°C; in solvent store at -80°C up to 6 months or -20°C for shorter term.