Learn More
Medchemexpress LLC Estradiol 3-methyl ether | 1035-77-4 | 99.9% | 286.41 | 10 MG
SDP
Catalog No. 5000452153
Encompass_Preferred
Estradiol 3-methyl ether is a synthetic estrogen that can disrupt microtubule networks in V79 cells with an EC50 of 9 μM and inhibit cell proliferation. Additionally, it decreases serum cholesterol levels and increases body weight loss, showing feminizing activity in an HFD murine model.
  • Synthetic estrogen
  • Disrupts microtubule networks
  • Inhibits cell proliferation
  • Decreases serum cholesterol levels
  • Increases body weight loss
  • Useful for biochemical assays
  • Potential for research into microtubule modulation and cell proliferation
  • Applicable in studies related to cholesterol regulation and weight management in relevant models
Catalog No. 50-004-52153 Supplier Medchemexpress LLC Supplier No. HYW00858110MG
Estradiol 3-methyl ether is a synthetic estrogen that can disrupt microtubule networks in V79 cells with an EC50 of 9 μM and inhibit cell proliferation. Additionally, it decreases serum cholesterol levels and increases body weight loss, showing feminizing activity in an HFD murine model.
  • Synthetic estrogen
  • Disrupts microtubule networks
  • Inhibits cell proliferation
  • Decreases serum cholesterol levels
  • Increases body weight loss
  • Useful for biochemical assays
  • Potential for research into microtubule modulation and cell proliferation
  • Applicable in studies related to cholesterol regulation and weight management in relevant models