Learn More
Medchemexpress LLC Estradiol 3-methyl ether | 1035-77-4 | 99.9% | 286.41 | 25 MG
SDP
Catalog No. 5000366721
Encompass_Preferred
Estradiol 3-methyl ether is a synthetic estrogen. It can disrupt microtubule networks in V79 cells with an EC50 of 9 μM and inhibits cell proliferation. Estradiol 3-methyl ether has also been shown to decrease serum cholesterol levels and increase body weight loss, indicating feminizing activity in an HFD murine model.
  • Synthetic estrogen
  • Disrupts microtubule networks in V79 cells (EC50 of 9 μM)
  • Inhibits cell proliferation
  • Decreases serum cholesterol levels
  • Increases body weight loss in HFD murine models
Catalog No. 50-003-66721 Supplier Medchemexpress LLC Supplier No. 25MGHYW008581
Estradiol 3-methyl ether is a synthetic estrogen. It can disrupt microtubule networks in V79 cells with an EC50 of 9 μM and inhibits cell proliferation. Estradiol 3-methyl ether has also been shown to decrease serum cholesterol levels and increase body weight loss, indicating feminizing activity in an HFD murine model.
  • Synthetic estrogen
  • Disrupts microtubule networks in V79 cells (EC50 of 9 μM)
  • Inhibits cell proliferation
  • Decreases serum cholesterol levels
  • Increases body weight loss in HFD murine models