Learn More
Medchemexpress LLC Estradiol 3-sulfamate | 172377-52-5 | 351.46 | 1 ML
SDP
Catalog No. 5000447054
Encompass_Preferred
Estradiol 3-sulfamate is a potent, long-acting, and orally active steroid sulfatase inhibitor. It effectively inhibits estrone sulfatase with specific IC50 and Ki values, making it suitable for various research applications.
  • Potent steroid sulfatase inhibitor
  • Long-acting steroid sulfatase inhibitor
  • Orally active
  • Inhibits estrone sulfatase with an IC50 of 251 nM
  • Inhibits estrone sulfatase with a Ki of 133 nM
  • For research use only
Catalog No. 50-004-47054 Supplier Medchemexpress LLC Supplier No. HYU0011210MM/1ML
Estradiol 3-sulfamate is a potent, long-acting, and orally active steroid sulfatase inhibitor. It effectively inhibits estrone sulfatase with specific IC50 and Ki values, making it suitable for various research applications.
  • Potent steroid sulfatase inhibitor
  • Long-acting steroid sulfatase inhibitor
  • Orally active
  • Inhibits estrone sulfatase with an IC50 of 251 nM
  • Inhibits estrone sulfatase with a Ki of 133 nM
  • For research use only