Learn More
Medchemexpress LLC Estradiol 3-sulfamate | 172377-52-5 | 351.46 | 5 MG
SDP
Catalog No. 5000446616
Encompass_Preferred
Estradiol 3-sulfamate is a potent, long-acting, and orally active steroid sulfatase inhibitor. It inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.
  • Potent, long-acting, and orally active steroid sulfatase inhibitor.
  • Inhibits estrone sulfatase.
  • Used for research purposes.
Catalog No. 50-004-46616 Supplier Medchemexpress LLC Supplier No. HYU001125MG
Estradiol 3-sulfamate is a potent, long-acting, and orally active steroid sulfatase inhibitor. It inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.
  • Potent, long-acting, and orally active steroid sulfatase inhibitor.
  • Inhibits estrone sulfatase.
  • Used for research purposes.