Learn More
Cayman Chemical Estradiol 3-sulfate 17β-Glucuronide (potassium salt), Purity: ≥95%, CAS: 10392-35-5, 5 mg
SDP
Catalog No. 502966785
Encompass_Preferred
A metabolite of estradiol 3-sulfate that has been modified by a UDP-glucuronosyltransferase; enhances utilization by anion transporters, most commonly leading to excretion in urine and bile
Catalog No. 50-296-6785 Supplier Cayman Chemical Supplier No. 182245 MG
A metabolite of estradiol 3-sulfate that has been modified by a UDP-glucuronosyltransferase; enhances utilization by anion transporters, most commonly leading to excretion in urine and bile