Learn More
Medchemexpress LLC HY-B0141 500mg , Estradiol CAS:50-28-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0141500MG
Encompass_Preferred
HY-B0141 500mg Estradiol CAS: 50-28-2 Estradiol (β-Estradiol) is a steroid hormone and the major female sex hormone. Estradiol can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. Estradiol can be used for the research of cancers, neurodegenerative diseases and neural tissue engineering. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9396