Learn More
Apexbio Technology LLC α-Estradiol 57-91-0 50mg
SDP
Catalog No. 5000088498
Encompass_Preferred
alpha-Estradiol (57-91-0) is a small-molecule inhibitor targeting 5 -reductase the enzyme responsible for converting testosterone to dihydrotestosterone (DHT) It is designed to inhibit 5 -reductase activity thereby modulating androgen metabolism and androgen-related signaling pathways alpha-Estradiol exerts its biological activity primarily through enzyme inhibition In reported experimental studies alpha-Estradiol demonstrates inhibitory activity against 5 -reductase with an IC50 value ranging from approximately 1 to 10 M though the precise value may depend on assay conditions Based on these pharmacological properties alpha-Estradiol holds research potential in the treatment and study of androgenetic alopecia
Catalog No. 50-000-88498 Supplier Apexbio Technology LLC Supplier No. B720150
alpha-Estradiol (57-91-0) is a small-molecule inhibitor targeting 5 -reductase the enzyme responsible for converting testosterone to dihydrotestosterone (DHT) It is designed to inhibit 5 -reductase activity thereby modulating androgen metabolism and androgen-related signaling pathways alpha-Estradiol exerts its biological activity primarily through enzyme inhibition In reported experimental studies alpha-Estradiol demonstrates inhibitory activity against 5 -reductase with an IC50 value ranging from approximately 1 to 10 M though the precise value may depend on assay conditions Based on these pharmacological properties alpha-Estradiol holds research potential in the treatment and study of androgenetic alopecia