Learn More
Medchemexpress LLC β-estradiol-6-one 6-(O-carboxymethyloxime) | 35048-47-6 | MFCD00056459 | 99.7% | 359.42 g/mol | C20H25NO5 | 5 MG
SDP
Catalog No. 5000262530
Encompass_Preferred
β-Estradiol-6-one 6-(O-carboxymethyloxime) is an estradiol-derived small molecule functionalized at the 6-position with an O-carboxymethyloxime moiety. It is used as an alkyl chain-based PROTAC linker and building block for synthesizing targeted protein degradation constructs. Provided for research use only.
  • Designed as an alkyl chain-based PROTAC linker for conjugation chemistry.
  • High purity suitable for analytical and synthetic applications.
  • Solid form, easy to handle and store.
  • Molecular formula C20H25NO5 and molecular weight 359.42 g/mol.
  • Available in small pack sizes for research and method development.
Catalog No. 50-002-62530 Supplier Medchemexpress LLC Supplier No. HY1334075MG
β-Estradiol-6-one 6-(O-carboxymethyloxime) is an estradiol-derived small molecule functionalized at the 6-position with an O-carboxymethyloxime moiety. It is used as an alkyl chain-based PROTAC linker and building block for synthesizing targeted protein degradation constructs. Provided for research use only.
  • Designed as an alkyl chain-based PROTAC linker for conjugation chemistry.
  • High purity suitable for analytical and synthetic applications.
  • Solid form, easy to handle and store.
  • Molecular formula C20H25NO5 and molecular weight 359.42 g/mol.
  • Available in small pack sizes for research and method development.