Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals β-Estradiol, 98%

Catalog No. AC436320250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436320250 25 g Glass bottle
AC436320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436320250 Supplier Thermo Scientific Chemicals Supplier No. 436320250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

β-Estradiol, 98%, CAS # 50-28-2, is an estrogen steroid sex hormone that regulates several biological functions including gene transcription and protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• β-Estradiol is a naturally occurring endogenous hormone in females

• It is an important steroid sex hormone involved in the regulation of several biological functions including gene transcription and protein synthesis

• This hormone is produced by the ovaries and can bind to and activate specific nuclear receptors

Applications

• β-Estradiol acts as an estrogenic hormone receptor agonist and activator of PI 3-kinase

• It shows inhibitory activity against aldehyde oxidase

• β-Estradiol presents mild anabolic and metabolic activities and increases blood coagulability in vivo experiments

• It plays a role in immune and inflammatory processes

• β-Estradiol triggers an increase in hepatic synthesis of some proteins, including sex hormone-binding globulin (SHBG), and thyroid-binding globulin (TBG), and suppresses the formation of follicle-stimulating hormone (FSH)

• This compound has been used for the in vitro maturation of bovine cumulus-oocyte complexes (COCs)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-28-2
Molecular Formula C18H24O2
Molecular Weight (g/mol) 272.38
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
PubChem CID 5757
ChEBI CHEBI:16469
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 176.0°C to 180.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,3758
Specific Rotation +76° to +83° (20°C, 589nm) (c=1, dioxane)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform, hexane, ethyl acetate
Formula Weight 272.38
Percent Purity 98%
Physical Form Powder
Chemical Name or Material β-Estradiol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause harm to breast-fed children.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
EINECSNumber 200-023-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.