Learn More
American Radiolabeled Chemicals Inc ESTRADIOL-B-D-GLU-3H 50UC
SDP
Catalog No. 5090104983
Encompass
Estradiol-6,7-3H(N)-17-beta-D-glucuronide 50uCi in ethanol:water (9:1), 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 448.5, Shipped in Dry Ice
Catalog No. 50-901-04983 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 132050UC
Estradiol-6,7-3H(N)-17-beta-D-glucuronide 50uCi in ethanol:water (9:1), 1 mCi/ml, S.A. 30-60 Ci/mmol, M.W. 448.5, Shipped in Dry Ice