Learn More
Selleck Chemical LLC Estradiol Benzoate S4110-50mg
SDP
Catalog No. 503111099
Encompass_Preferred
Estradiol benzoate( -Estradiol 3-benzoate) is an estradiol analog which binds to the human murine and chicken ER with IC50 of 22-28 nM
Catalog No. 50-311-1099 Supplier Selleck Chemical LLC Supplier No. S411050MG
Add to Cart
Add to Cart
Estradiol benzoate( -Estradiol 3-benzoate) is an estradiol analog which binds to the human murine and chicken ER with IC50 of 22-28 nM