Learn More
U.S. Pharmacopeia Estradiol Benzoate, 50-50-0, MFCD00003692, 250mg
Supplier:  U.S. Pharmacopeia 1251000
Encompass
Molecular formula C25H28O3, Molecular weight 376.49, Melting Point 375.8 - 386.6 °F (191 - 197 °C), Synonyms: 17-beta-Estradiol 3-benzoate
Catalog No. NC1503449