Learn More
Medchemexpress LLC Estradiol benzoate | 50-50-0 | 1 G
SDP
Supplier:  Medchemexpress LLC HYB11921G
Encompass_Preferred
Estradiol benzoate (β-Estradiol 3-benzoate) is an HBx protein inhibitor that inhibits androgen and hepatitis B virus (HBV) transcription and replication. It also shows antifertility effects, anti-Toxoplasma gondii activity, and can improve memory behavior in Ovariectomy (Ovx) female mice.
  • Inhibits HBx protein.
  • Suppresses androgen and hepatitis B virus (HBV) transcription and replication.
  • Exhibits antifertility effects.
  • Shows anti-Toxoplasma gondii activity.
  • Improves memory behavior in ovariectomy (Ovx) female mice.
  • Reverses lowered uterine weight in Ovx mice.
  • Increases number and density of synaptic vesicles.
Catalog No. 50-004-13597