Learn More
Medchemexpress LLC Estradiol benzoate | 50-50-0 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB119210MM/1ML
Encompass_Preferred
Estradiol benzoate is identified as an HBx protein inhibitor that effectively suppresses both androgen and hepatitis B virus (HBV) transcription and replication. Additionally, it demonstrates antifertility properties, exhibits activity against Toxoplasma gondii, and has been observed to enhance memory behavior in ovariectomized female mice. This substance is strictly designated for research applications.
  • Acts as an HBx protein inhibitor.
  • Inhibits androgen transcription and replication.
  • Inhibits hepatitis B virus (HBV) transcription and replication.
  • Shows antifertility effects.
  • Exhibits anti-Toxoplasma gondii activity.
  • Can improve memory behavior in ovariectomy female mice.
Catalog No. 50-004-13598