Learn More
Medchemexpress LLC Estradiol benzoate | 50-50-0 | 99.9% | 250 MG
Supplier: Medchemexpress LLC 250MGHYB1192R
Estradiol benzoate is a HBx protein inhibitor that inhibits androgen and hepatitis B virus (HBV) transcription and replication. It also shows antifertility effects, anti-Toxoplasma gondii activity, and can improve memory behavior in Ovariectomy (Ovx) female mice.
- Inhibits HBx protein
- Inhibits androgen transcription
- Inhibits hepatitis B virus (HBV) transcription and replication
- Exhibits antifertility effects
- Shows anti-Toxoplasma gondii activity
- May improve memory behavior in Ovariectomy (Ovx) female mice