Learn More
Cayman Chemical Estradiol Benzoate
SDP
Supplier:  Cayman Chemical 100064871 G
Encompass_Preferred
An estradiol analog; often used in combination with a progestin to induce estrus in domestic livestock; binds to human, murine, and chicken ERα IC50s = 22-28 nM)
Catalog No. 50-288-8089