Learn More
Cayman Chemical Estradiol Benzoate, Purity: ≥98%, CAS: 50-50-0, 500 mg
SDP
Catalog No. 502888090
Encompass_Preferred
An estradiol analog; often used in combination with a progestin to induce estrus in domestic livestock; binds to human, murine, and chicken ERα IC50s = 22-28 nM)
Catalog No. 50-288-8090 Supplier Cayman Chemical Supplier No. 10006487500 MG
An estradiol analog; often used in combination with a progestin to induce estrus in domestic livestock; binds to human, murine, and chicken ERα IC50s = 22-28 nM)