Learn More
Cayman Chemical Estradiol Benzoate, Purity: ≥98%, CAS: 50-50-0, 5 g
SDP
Catalog No. 502891415
Encompass_Preferred
An estradiol analog; often used in combination with a progestin to induce estrus in domestic livestock; binds to human, murine, and chicken ERα IC50s = 22-28 nM)
Catalog No. 50-289-1415 Supplier Cayman Chemical Supplier No. 100064875 G
An estradiol analog; often used in combination with a progestin to induce estrus in domestic livestock; binds to human, murine, and chicken ERα IC50s = 22-28 nM)