Learn More
Selleck Chemical LLC Estradiol Benzoate S4110-10mM/1mL
SDP
Catalog No. 503108615
Encompass_Preferred
Estradiol benzoate( -Estradiol 3-benzoate) is an estradiol analog which binds to the human murine and chicken ER with IC50 of 22-28 nM
Catalog No. 50-310-8615 Supplier Selleck Chemical LLC Supplier No. S411010MM/1ML
Estradiol benzoate( -Estradiol 3-benzoate) is an estradiol analog which binds to the human murine and chicken ER with IC50 of 22-28 nM